Learn More
TARGETMOL CHEMICALS INC TRI-GALNAC-COOH 5MG
SDP
Catalog No. 502580056
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tri-GalNAc-COOH is a ligand for the asialoglycoprotein receptor (ASGPR) commonly employed in Lysosome Targeting Chimera (LYTAC) investigations. purity: 99%
Catalog No. 50-258-0056 Supplier TARGETMOL CHEMICALS INC Supplier No. T393705MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tri-GalNAc-COOH is a ligand for the asialoglycoprotein receptor (ASGPR) commonly employed in Lysosome Targeting Chimera (LYTAC) investigations. purity: 99%