Learn More
Medchemexpress LLC Tri-GalNAc-DBCO | 2765060-22-6 | 96.6% | 1950.18 | C91H144N12O34 | 10 MG
SDP
Catalog No. 5000285489
Encompass_Preferred
Trivalent GalNAc-DBCO is a synthetic trivalent N-acetylgalactosamine (GalNAc) ligand functionalized with a dibenzocyclooctyne (DBCO) moiety. It is intended for strain-promoted alkyne-azide cycloaddition (SPAAC) to enable copper-free click conjugation to azide-modified oligonucleotides and other biomolecules, and can direct conjugated cargo to the asialoglycoprotein receptor (ASGPR) for hepatocyte targeting and lysosomal trafficking.
  • Enables copper-free click conjugation via strain-promoted alkyne-azide cycloaddition.
  • Designed for conjugation to azide-modified oligonucleotides and biomolecules.
  • Triantennary GalNAc motif targets ASGPR for liver-directed delivery.
  • High purity suitable for biochemical and conjugation workflows.
  • Available in small-scale 10 mg packs for research use.
Catalog No. 50-002-85489 Supplier Medchemexpress LLC Supplier No. HY16029110MG
Trivalent GalNAc-DBCO is a synthetic trivalent N-acetylgalactosamine (GalNAc) ligand functionalized with a dibenzocyclooctyne (DBCO) moiety. It is intended for strain-promoted alkyne-azide cycloaddition (SPAAC) to enable copper-free click conjugation to azide-modified oligonucleotides and other biomolecules, and can direct conjugated cargo to the asialoglycoprotein receptor (ASGPR) for hepatocyte targeting and lysosomal trafficking.
  • Enables copper-free click conjugation via strain-promoted alkyne-azide cycloaddition.
  • Designed for conjugation to azide-modified oligonucleotides and biomolecules.
  • Triantennary GalNAc motif targets ASGPR for liver-directed delivery.
  • High purity suitable for biochemical and conjugation workflows.
  • Available in small-scale 10 mg packs for research use.