Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC tri(GalNAc-PEG2)-PEG4-cyclo(Ac-DCAW-{Arg(3F-Ph)-PEG2}-LGELVWCT) (Cys2-Cys12) | 99.5% | 3488.76 Da | C152H236FN25O62S2 | 500 UG
SDP

Catalog No. 5000202748
Encompass_Preferred

tri(GalNAc-PEG2)-PEG4-cyclo(Ac-DCAW-{Arg(3F-Ph)-PEG2}-LGELVWCT) (Cys2-Cys12) is a cyclic GalNAc-conjugated peptide designed as an asialoglycoprotein receptor (ASGPR) ligand for lysosome-targeting chimera (LYTAC) research. It is supplied as a purified synthetic peptide in solid form for research use only.

  • Cyclic GalNAc-conjugated peptide designed to target ASGPR.
  • Suitable for lysosome-targeting chimera (LYTAC) research.
  • Molecular weight approximately 3488.76 Da.
  • Chemical formula C152H236FN25O62S2.
  • High reported purity (≈99.5%).
  • Supplied as a solid; recommended storage at -20°C, protected from light and under inert gas; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small pack sizes suitable for research (for example, 500 μg and up).

Catalog No. 50-002-02748 Supplier Medchemexpress LLC Supplier No. HY171910500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

tri(GalNAc-PEG2)-PEG4-cyclo(Ac-DCAW-{Arg(3F-Ph)-PEG2}-LGELVWCT) (Cys2-Cys12) is a cyclic GalNAc-conjugated peptide designed as an asialoglycoprotein receptor (ASGPR) ligand for lysosome-targeting chimera (LYTAC) research. It is supplied as a purified synthetic peptide in solid form for research use only.

  • Cyclic GalNAc-conjugated peptide designed to target ASGPR.
  • Suitable for lysosome-targeting chimera (LYTAC) research.
  • Molecular weight approximately 3488.76 Da.
  • Chemical formula C152H236FN25O62S2.
  • High reported purity (≈99.5%).
  • Supplied as a solid; recommended storage at -20°C, protected from light and under inert gas; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small pack sizes suitable for research (for example, 500 μg and up).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.