Learn More
Strem, An Ascensus Company CAS# 6476-36-4. 1g. Tri-i-propylphosphine, 98%. MFCD00015029
SDP
Catalog No. 5090110476
Encompass
CAS# 6476-36-4. 1g. Tri-i-propylphosphine, 98%. MFCD00015029. Molecular Weight: 160.24. Molecular Formula: (i-C3H7)3P. Color/form: colorless liq. Strem# 15-6950. http://www.strem.com/catalog/v/15-6950/
Catalog No. 50-901-10476 Supplier Strem, An Ascensus Company Supplier No. 1569501G
CAS# 6476-36-4. 1g. Tri-i-propylphosphine, 98%. MFCD00015029. Molecular Weight: 160.24. Molecular Formula: (i-C3H7)3P. Color/form: colorless liq. Strem# 15-6950. http://www.strem.com/catalog/v/15-6950/