Learn More
Strem, An Ascensus Company CAS# 116-17-6. 50g. Tri-i-propylphosphite, min. 94%. MFCD00008874
SDP
Catalog No. 5090110483
Encompass
CAS# 116-17-6. 50g. Tri-i-propylphosphite, min. 94%. MFCD00008874. Molecular Weight: 208.24. Molecular Formula: [(CH3)2CHO]3P. Color/form: colorless liq. Strem# 15-7000. http://www.strem.com/catalog/v/15-7000/
Catalog No. 50-901-10483 Supplier Strem, An Ascensus Company Supplier No. 15700050G
CAS# 116-17-6. 50g. Tri-i-propylphosphite, min. 94%. MFCD00008874. Molecular Weight: 208.24. Molecular Formula: [(CH3)2CHO]3P. Color/form: colorless liq. Strem# 15-7000. http://www.strem.com/catalog/v/15-7000/