Learn More
Cusabio Technology LLC Rat Tri-iodothyronine,T3 ELISA Kit
Supplier:  Cusabio Technology LLC CSBE05085R
Encompass_Preferred
This Rat Tri-iodothyronine (T3) ELISA Kit was designed for the quantitative measurement of Rat Tri-iodothyronine (T3) protein in serum, plasma, cell culture supernates, tissue homogenates. It is a Competitive ELISA kit, its detection range is 0.5 ng/mL-8 ng/mL and the sensitivity is 0.5 ng/mL. One kit contains 96 wells.
Catalog No. NC2952846