Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Tri-level Beta-hydroxybutyrate (BHB) Controls
SDP

Catalog No. 23001178 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
6 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-001-178 6 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-178
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Fisherbrand Beta-hydroxybutyrate (BHB) Tri-level controls kit can be used to provide quality control of the Beta-hydroxybutyrate (BHB) Liquicolor™ reagent kit.

  • The Tri-level Control Solution for Beta-Hydroxybutyrate includes accurate measurements of Acetaminophen, Beta-Hydroxybutyrate, and Salicylate to ensure dependable testing.
  • Ready-to-use liquid controls for immediate use, saving time and effort.
  • Compatible with Serum or Plasma: Suitable for use with serum or plasma samples, providing flexibility in sample selection.
  • Linear Range up to 4.5 mmol/L: Offers a linear measurement range of up to 4.5 mmol/L for precise results.
  • 60-Day Open Vial Stability: Stable for 60 days after opening when refrigerated (2 to 8°C), minimizing waste.
  • FDA Cleared
  • Matrix-Free Serum Substitute: Utilizes a synthetic serum substitute (SeraSub™) matrix, free of serum infectious agents.

Specifications

Certifications/Compliance MDSAP, ISO13485:2016, FDA-Cleared
Content And Storage Refrigerated
Concentration Dependent
Detectable Analytes beta-hydroxybutyrate (BHB), Acetaminophen, Salicylate
For Use With (Equipment) Any open channel analyzer
Type TDM/Beta-Hydroxybutyrate Tri-Level Control
Disposable Reusable
Includes Three levels of unassayed controls with concentrations of analytes Acetaminophen, Beta-Hydroxybutyrate, and Salicylate.
Level 3
Quantity 6 x 5 mL
Stability 60 days after opening at 2°C to 8°C
Sterility Non-sterile
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.