Learn More
Cell Signaling Technology Matched Antibody Pair, Tri-Methyl-Histone H3 (Lys27), Unconjugated, Human, Mouse, Rat, Monkey, 1 Kit
SDP
Catalog No. 503133923
Encompass
The Tri-Methyl-Histone H3 (Lys27) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling, such as biotin, beads, fluorophores, or lanthanides. This kit is suitable for use with platforms including MSD, Quanterix Simoa, Luminex, and Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF).
  • Includes capture and detection antibodies that bind to non-overlapping epitopes
  • Supplied as BSA and azide free formulations
  • Validated for human, mouse, rat, and monkey reactivity
  • Compatible with high-throughput ELISA and immunoassay platforms
  • Consists of two rabbit monoclonal antibodies
Catalog No. 50-313-3923 Supplier Cell Signaling Technology Supplier No. 92075S
The Tri-Methyl-Histone H3 (Lys27) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require antibody pairs with specialized or custom labeling, such as biotin, beads, fluorophores, or lanthanides. This kit is suitable for use with platforms including MSD, Quanterix Simoa, Luminex, and Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF).
  • Includes capture and detection antibodies that bind to non-overlapping epitopes
  • Supplied as BSA and azide free formulations
  • Validated for human, mouse, rat, and monkey reactivity
  • Compatible with high-throughput ELISA and immunoassay platforms
  • Consists of two rabbit monoclonal antibodies