Learn More
BIOHIPPO Tri-Methyllysine pAb
SDP
Catalog No. 5000975503
Encompass
U-Blot Pan-Tri-Methyllysine Rabbit pAb (KLH-conjugate immunogen) is a Rabbit-derived polyclonal antibody that specifically recognizes Tri-Methyllysine a lysine trimethylation modification at Lys(K) It shows reactivity with Species Independent Validated applications include ELISA WB IP IF IAC Recommended dilutions ELISA 12000 WB 12000 IP 5 ug/mg protein sample IF 5 ug/mL or 1100 Purified by n-(epsilon) trimethyllysine on agarose and immunoaffinity absorbed with n-(epsilon) methylated lysine and n-(epsilon) dimethylated lysine on agarose Histones undergo a wide range of enzyme-catalyzed post-translational modifications including acetylation methylation phosphorylation and ubiquitination Lysine trimethylation is a vital epigenetic modification that predominantly occurs on histone lysine residues It is dynamically regulated by histone methyltransferases and demethylases and extensively distributed in eukaryotic chromatin Supplied as 100 uL
Catalog No. 50-009-75503 Supplier BIOHIPPO Supplier No. BHA21821428
U-Blot Pan-Tri-Methyllysine Rabbit pAb (KLH-conjugate immunogen) is a Rabbit-derived polyclonal antibody that specifically recognizes Tri-Methyllysine a lysine trimethylation modification at Lys(K) It shows reactivity with Species Independent Validated applications include ELISA WB IP IF IAC Recommended dilutions ELISA 12000 WB 12000 IP 5 ug/mg protein sample IF 5 ug/mL or 1100 Purified by n-(epsilon) trimethyllysine on agarose and immunoaffinity absorbed with n-(epsilon) methylated lysine and n-(epsilon) dimethylated lysine on agarose Histones undergo a wide range of enzyme-catalyzed post-translational modifications including acetylation methylation phosphorylation and ubiquitination Lysine trimethylation is a vital epigenetic modification that predominantly occurs on histone lysine residues It is dynamically regulated by histone methyltransferases and demethylases and extensively distributed in eukaryotic chromatin Supplied as 100 uL