Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-butyl(vinyl)tin, 96%

Catalog No. AAL2021503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2021503 1 g
AAL2021506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2021503 Supplier Thermo Scientific Chemicals Supplier No. L2021503
Add to Cart

CAS: 7486-35-3 | C14H30Sn | 317.104 g/mol

Tributyl(vinyl)tin is used as a reagent in the palladium catalyzed synthesis of allyl and benzyl ethers. It is also a vinyl nucleophile for bromoacetylene and bromoaromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tributyl(vinyl)tin is used as a reagent in the palladium catalyzed synthesis of allyl and benzyl ethers. It is also a vinyl nucleophile for bromoacetylene and bromoaromatics.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7486-35-3
Molecular Formula C14H30Sn
MDL Number MFCD00009421
InChI Key QIWRFOJWQSSRJZ-UHFFFAOYSA-N
Synonym tributyl vinyl tin, vinyltributyltin, vinyltributylstannane, tributyl vinyl stannane, tributyl ethenyl stannane, stannane, tributylethenyl, vinyltributyl tin, stannane, tributylvinyl, tributylstannylethylene, vinyltri-n-butyltin
PubChem CID 81998
IUPAC Name tributyl(ethenyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C=C

Specifications

Density 1.085
Boiling Point 104°C to 106°C (3.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.478
Quantity 1 g
UN Number UN2788
Beilstein 3537662
Sensitivity Moisture sensitive
Solubility Information Not miscible or difficult to mix in water.
Molecular Weight (g/mol) 317.104
Formula Weight 317.11
Percent Purity 96%
Chemical Name or Material Tri-n-butyl(vinyl)tin
Show More Show Less
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
EINECSNumber 231-291-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.