Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tri-N-Butylphosphine

Catalog No. T4948425ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T4948425ML 25 mL
T494845ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. T4948425ML Supplier Sigma Aldrich Supplier No. T4948425ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,4-addition catalyst; used with disulfides for the thioetherification of alcohols; acylation catalyst; used to prepare active esters.

Specifications

CAS 1461-22-9
Density 1.2 g/mL at 25°C
Boiling Point 171°C to 173°C (25 mmHg, lit.)
Molecular Formula C12H27ClSn
Linear Formula [CH3(CH2)3]3SnCl
Refractive Index n20/D 1.462
MDL Number MFCD00000521
Quantity 25 mL
Synonym Tributylphosphine; NSC 91700; P(n-Bu)3; TBP
Molecular Weight (g/mol) 325.51 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.