Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tri-N-Butylphosphine

Catalog No. 731374100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-137-4100G 100 g
73-137-425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 73-137-4100G Supplier Sigma Aldrich Supplier No. 731374100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Catalyst for:Domino reactions of activated conjugated dienes with +a,+a-unsaturated-+a-ketoesters[3+2]-CycloadditionsUmpolung addition reactionsReductive carbonylationAllylation reactions

Specifications

CAS 998-40-3
Density 0.81 g/mL at 25°C
Boiling Point 150°C (50 mmHg, lit.)
Molecular Formula C12H27P
Refractive Index n20/D 1.462
Linear Formula [CH3(CH2)3]3P
MDL Number MFCD00009462
Quantity 100 g
Synonym Tributylphosphine; NSC 91700; P(n-Bu)3; TBP
Molecular Weight (g/mol) 202.32 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.