Learn More
Strem, An Ascensus Company CAS# 998-40-3. 250g. Tri-n-butylphosphine, 99% (10 wt% in hexanes). MFCD00009462
SDP
Catalog No. 5090110363
Encompass
CAS# 998-40-3. 250g. Tri-n-butylphosphine, 99% (10 wt% in hexanes). MFCD00009462. Molecular Weight: 202.32. Molecular Formula: (n-C4H9)3P. Color/form: colorless liq. Strem# 15-5802. http://www.strem.com/catalog/v/15-5802/
Catalog No. 50-901-10363 Supplier Strem, An Ascensus Company Supplier No. 155802250G
CAS# 998-40-3. 250g. Tri-n-butylphosphine, 99% (10 wt% in hexanes). MFCD00009462. Molecular Weight: 202.32. Molecular Formula: (n-C4H9)3P. Color/form: colorless liq. Strem# 15-5802. http://www.strem.com/catalog/v/15-5802/