Learn More
Strem, An Ascensus Company CAS# 998-40-3. 100g. Tri-n-butylphosphine, min. 93%. MFCD00009462
SDP
Catalog No. 5090110361
Encompass
CAS# 998-40-3. 100g. Tri-n-butylphosphine, min. 93%. MFCD00009462. Molecular Weight: 202.32. Molecular Formula: (n-C4H9)3P. Color/form: colorless to light yellow liq. Strem# 15-5801. http://www.strem.com/catalog/v/15-5801/
Catalog No. 50-901-10361 Supplier Strem, An Ascensus Company Supplier No. 155801100G
CAS# 998-40-3. 100g. Tri-n-butylphosphine, min. 93%. MFCD00009462. Molecular Weight: 202.32. Molecular Formula: (n-C4H9)3P. Color/form: colorless to light yellow liq. Strem# 15-5801. http://www.strem.com/catalog/v/15-5801/