Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-butyltin fluoride

Catalog No. AA3960206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3960206 5 g
AA3960214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3960206 Supplier Thermo Scientific Chemicals Supplier No. 03960206
Add to Cart

CAS: 1983-10-4 | C8H15NaO2 | 166.20 g/mol

Tri-n-butyltin fluoride is a reagent used for the selective desilylation of bis(silyl) enol ethers. It is also used as antifoulant for marine paints; reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1983-10-4
MDL Number MFCD00013547

Specifications

Melting Point 248°C to 252°C
Odor Pungent
Molecular Formula C12H29FSn
Quantity 5 g
UN Number UN3146
Sensitivity Moisture sensitive
Synonym fluorotributyltin, tributyltin fluoride, stannane, tributylfluoro, fluorotributylstannane, tri-n-butyltin fluoride, stannane, fluorotributyl, fluorotri-n-butyltin, tin, tributylfluoro, tributylstannane fluoride, tri-n-butylstannyl fluoride
Solubility Information Soluble in nonpolar organic solvents. Insoluble in water.
InChI Key STUMXSORVXBBJR-UHFFFAOYSA-N
SMILES CCCC[Sn](CCCC)(CCCC)F
IUPAC Name tributyl(fluoro)stannane
Molecular Weight (g/mol) 311.07
PubChem CID 16120
Formula Weight 309.04
Physical Form Powder
Chemical Name or Material Tri-n-butyltin fluoride
Show More Show Less
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECSNumber 217-847-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.