Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-N-Caprin, ∽99%, MP Biomedicals™

Catalog No. ICN10529701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10529701 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10529701 Supplier MP Biomedicals, Inc Supplier No. 0210529701
Add to Cart
Add to Cart

Chemical Identifiers

CAS 621-71-6
Molecular Formula C33H62O6
Molecular Weight (g/mol) 554.853
InChI Key LADGBHLMCUINGV-UHFFFAOYSA-N
Synonym tricaprin, tridecanoin, glycerol tricaprate, glycerol tridecanoate, decanoin, tri, tricapric glyceride, glycerol tricaprin, 1,2,3-tridecanoylglycerol, capric acid triglyceride, glyceryl tricaprate
PubChem CID 69310
ChEBI CHEBI:77388
IUPAC Name 2,3-di(decanoyloxy)propyl decanoate
SMILES CCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCC)OC(=O)CCCCCCCCC

Specifications

Color Yellow
Assay Percent Range ∽99%
Quantity 1 g
Formula Weight 554.8
Physical Form Solid
Chemical Name or Material Tri-N-Caprin
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.