Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-decylamine 97.0+%, TCI America™
SDP

Catalog No. T041825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T041825G 25 g
T04185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T041825G Supplier TCI America Supplier No. T041825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1070-01-5
Molecular Formula C30H63N
Molecular Weight (g/mol) 437.841
MDL Number MFCD00041914
InChI Key COFKFSSWMQHKMD-UHFFFAOYSA-N
Synonym tri-n-decylamine, tris decyl amine, 1-decanamine, n,n-didecyl, tridecanitrile mixed isomers, trisdecylamine, tri decyl amine, n,n-didecyl-1-decanamine, 4-04-00-00785 beilstein handbook reference, tri-n-decylamine tertiary amine
PubChem CID 14049
IUPAC Name N,N-didecyldecan-1-amine
SMILES CCCCCCCCCCN(CCCCCCCCCC)CCCCCCCCCC

Specifications

Color Yellow
Boiling Point 430°C
Quantity 25 g
Formula Weight 437.84
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Tri-n-decylamine
EINECSNumber (2)-0139
RTECSNumber YD3675000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.