Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-octyl Borate 98.0+%, TCI America™
SDP

Catalog No. B209525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B209525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B209525G Supplier TCI America Supplier No. B209525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2467-12-1
Molecular Formula C24H51BO3
Molecular Weight (g/mol) 398.48
MDL Number MFCD00027324
InChI Key DTBRTYHFHGNZFX-UHFFFAOYSA-N
Synonym Boric Acid Tri-n-octyl Ester
PubChem CID 17176
IUPAC Name trioctyl borate
SMILES B(OCCCCCCCC)(OCCCCCCCC)OCCCCCCCC

Specifications

Color Yellow
Boiling Point 379°C
Quantity 25 g
Formula Weight 398.48
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Tri-n-octyl Borate
RTECSNumber ED5630000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.