Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-o-tolyl Borate 98.0+%, TCI America™
SDP

Catalog No. B05195ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B05195ML 5 mL
B051925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B05195ML Supplier TCI America Supplier No. B05195ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2665-12-5
Molecular Formula C21H21BO3
Molecular Weight (g/mol) 332.21
MDL Number MFCD00059311
InChI Key RTMBXAOPKJNOGZ-UHFFFAOYSA-N
Synonym Boric Acid Tri-o-cresyl Ester, Tri-o-cresyl Borate, Boric Acid Tri-o-tolyl Ester
PubChem CID 75875
IUPAC Name tris(2-methylphenyl) borate
SMILES B(OC1=CC=CC=C1C)(OC2=CC=CC=C2C)OC3=CC=CC=C3C

Specifications

Color Yellow
Boiling Point 386°C
Quantity 5 mL
Formula Weight 332.21
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Tri-o-tolyl Borate
RTECSNumber ED4960000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.