Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-o-tolyl phosphate, Analytical Standard, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111019706 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9706 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9706 Supplier MilliporeSigma Supelco Supplier No. 51885250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tri-o-tolyl phosphate is classified under the class of non-halogenated organophosphate esters (OPEs).

TRUSTED_SUSTAINABILITY

Specifications

CAS 78-30-8
Linear Formula C21H21O4P
MDL Number MFCD00041906
Synonym NSC 438; Phosphoric acid tris(2-methylphenyl) ester; TOTP; Tri-o-cresyl phosphate
Quantity 250 mg
Formula Weight 368.36
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.