Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-o-tolyl Phosphite 97.0+%, TCI America™
SDP

Catalog No. P14165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14165G 5 g
P14161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14165G Supplier TCI America Supplier No. P14165G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2622-08-4
Molecular Formula C21H21O3P
Molecular Weight (g/mol) 352.37
MDL Number MFCD00014911
InChI Key BKHZQJRTFNFCTG-UHFFFAOYSA-N
Synonym tri-o-tolyl phosphite, tri-o-tolylphosphite, tri-o-cresyl phosphite, phosphorous acid tri-o-cresyl ester, tris o-methylphenyl phosphite, tris 2-methylphenyl phosphite, tris 2-tolyl phosphite, tris o-tolyloxy phosphine, phosphorous acid, tri-o-cresyl ester, tri-2-tolylphosphite
PubChem CID 92937
IUPAC Name tris(2-methylphenyl) phosphite
SMILES CC1=CC=CC=C1OP(OC1=CC=CC=C1C)OC1=CC=CC=C1C

Specifications

Color Yellow
Boiling Point 238°C
Quantity 5 g
Formula Weight 352.37
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tri-o-tolyl Phosphite
EINECSNumber (3)-2510
RTECSNumber TH1050000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.