Learn More
Strem, An Ascensus Company CAS# 69861-71-8. Bis(tri-o-tolylphosphine)palladium(0), min. 98%. MFCD12911908. 5G
SDP
Supplier:  Strem, An Ascensus Company 4602625G
Encompass
CAS# 69861-71-8. 5G. Bis(tri-o-tolylphosphine)palladium(0), min. 98%. MFCD12911908. Molecular Formula: C42H42P2Pd. Molecular Weight: 715.15. Color/Form: yellow xtl. Stability: air sensitive, store cold. Strem#: 46-0262 www.strem.com/catalog/v3/46-0262/
Catalog No. 50-195-9569