Learn More
Medchemexpress LLC Tri-(p-tert-butylphenyl) phosphate | 78-33-1 | 98.7% | 494.60 g·mol⁻¹ | C30H39O4P | 5 MG
SDP
Catalog No. 5000300129
Encompass_Preferred
Tri-(p-tert-butylphenyl) phosphate is an organophosphate compound used as a flame retardant and plasticizer in polymers and materials. It is identified by CAS 78-33-1 and is supplied for research applications.
  • Used as a flame retardant and plasticizer for polymers and materials.
  • Organophosphate chemical with formula C30H39O4P and molecular weight 494.60 g·mol⁻¹.
  • High purity: 98.69% (LCMS).
  • Storage: stable at room temperature for up to 3 years; in solvent: -80°C for 2 years, -20°C for 1 year.
  • Provided for research use applications.
Catalog No. 50-003-00129 Supplier Medchemexpress LLC Supplier No. HYW0199945G
Tri-(p-tert-butylphenyl) phosphate is an organophosphate compound used as a flame retardant and plasticizer in polymers and materials. It is identified by CAS 78-33-1 and is supplied for research applications.
  • Used as a flame retardant and plasticizer for polymers and materials.
  • Organophosphate chemical with formula C30H39O4P and molecular weight 494.60 g·mol⁻¹.
  • High purity: 98.69% (LCMS).
  • Storage: stable at room temperature for up to 3 years; in solvent: -80°C for 2 years, -20°C for 1 year.
  • Provided for research use applications.