Learn More
Medchemexpress LLC Tri-p-tolyl phosphate-d21 | 2469244-63-9 | MFCD00041908 | >98.0% | 389.49 g/mol | C21D21O4P | 2.5 MG
SDP
Catalog No. 5000284617
Encompass_Preferred
Tri-p-tolyl phosphate-d21 is the deuterium-labeled form of tri-p-tolyl phosphate supplied for research use as a tracer and internal standard in quantitative NMR, GC-MS, and LC-MS analyses. It is provided in small, pre-weighed quantities and should be stored according to the Certificate of Analysis; not for clinical or diagnostic use.
  • Deuterium-labeled internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, and LC-MS applications
  • Molecular formula C21D21O4P; molecular weight 389.49 g/mol
  • Available in pre-weighed quantities (e.g., 1 mg, 2.5 mg)
  • Store and handle per Certificate of Analysis; room temperature shipping may apply
Catalog No. 50-002-84617 Supplier Medchemexpress LLC Supplier No. HYW6543192.5MG
Tri-p-tolyl phosphate-d21 is the deuterium-labeled form of tri-p-tolyl phosphate supplied for research use as a tracer and internal standard in quantitative NMR, GC-MS, and LC-MS analyses. It is provided in small, pre-weighed quantities and should be stored according to the Certificate of Analysis; not for clinical or diagnostic use.
  • Deuterium-labeled internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, and LC-MS applications
  • Molecular formula C21D21O4P; molecular weight 389.49 g/mol
  • Available in pre-weighed quantities (e.g., 1 mg, 2.5 mg)
  • Store and handle per Certificate of Analysis; room temperature shipping may apply