Learn More
Medchemexpress LLC Tri-p-tolylamine-d21 | 201944-90-3 | 98.2% | 308.53 | 10 MG
SDP
Catalog No. 5000366950
Encompass_Preferred
Tri-p-tolylamine-d21 is the deuterium labeled Tri-p-tolylamine. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Isotope-labeled compounds
  • For research use only.
Catalog No. 50-003-66950 Supplier Medchemexpress LLC Supplier No. HYW009296S10MG
Tri-p-tolylamine-d21 is the deuterium labeled Tri-p-tolylamine. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Isotope-labeled compounds
  • For research use only.