Learn More
Medchemexpress LLC Tri-p-tolylamine-d21 | 201944-90-3 | 98.2% | 308.53 | 5 MG
SDP
Catalog No. 5000451561
Encompass_Preferred
Tri-p-tolylamine-d21 is the deuterium labeled Tri-p-tolylamine. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-51561 Supplier Medchemexpress LLC Supplier No. HYW009296S5MG
Tri-p-tolylamine-d21 is the deuterium labeled Tri-p-tolylamine. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS