Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tri-p-tolylphosphine | 1038-95-5 | ≥98.0% | 304.37 | 25 G
SDP

Catalog No. 5000366974
Encompass_Preferred

Tri-p-tolylphosphine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients. It is also used as a pharmaceutical intermediate.

  • Appearance is solid.
  • Color is white to off-white.
  • Shipping is at room temperature in continental US; may vary elsewhere.
  • Storage is at 4°C, stored under nitrogen. In solvent: -80°C for 6 months; -20°C for 1 month (stored under nitrogen).

Catalog No. 50-003-66974 Supplier Medchemexpress LLC Supplier No. 25GHYW011383
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tri-p-tolylphosphine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients. It is also used as a pharmaceutical intermediate.

  • Appearance is solid.
  • Color is white to off-white.
  • Shipping is at room temperature in continental US; may vary elsewhere.
  • Storage is at 4°C, stored under nitrogen. In solvent: -80°C for 6 months; -20°C for 1 month (stored under nitrogen).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.