Learn More
Strem, An Ascensus Company CAS# 1038-95-5. 5g. Tri-p-tolylphosphine, 98%. MFCD00008542
SDP
Catalog No. 5090110546
Encompass
CAS# 1038-95-5. 5g. Tri-p-tolylphosphine, 98%. MFCD00008542. Molecular Weight: 304.37. Molecular Formula: (p-CH3C6H4)3P. Color/form: white xtl. Strem# 15-8120. http://www.strem.com/catalog/v/15-8120/
Catalog No. 50-901-10546 Supplier Strem, An Ascensus Company Supplier No. 1581205G
CAS# 1038-95-5. 5g. Tri-p-tolylphosphine, 98%. MFCD00008542. Molecular Weight: 304.37. Molecular Formula: (p-CH3C6H4)3P. Color/form: white xtl. Strem# 15-8120. http://www.strem.com/catalog/v/15-8120/