Learn More
Chemglass Life Sciences 36" Tri Port Assembly, AdvantaSil APST Platinum Cured Silicone Tubing, 1/16" ID x 3/16" OD Size 14, PK3
SDP
Catalog No. 5000083166
Encompass
Features/Benefits: Single Use: Minimize Labor CostsEliminates Cleaning ValidationReduces Set up Time and Costs.Pre-Fabricated Assemblies: Minimize Injuries associated with cutting in-houseImproved consistency.Offers Closed System: Gives the ability to assemble in Open Lab type EnvironmentProvides aseptic solution to close off an unused line.How to Sterilize: Autoclave Max 15 psi @ 134 degreesC, 60 minGamma Radiation 25 kGy to 50 kGy.
Catalog No. 50-000-83166 Supplier Chemglass Life Sciences Supplier No. CLS139015
Features/Benefits: Single Use: Minimize Labor CostsEliminates Cleaning ValidationReduces Set up Time and Costs.Pre-Fabricated Assemblies: Minimize Injuries associated with cutting in-houseImproved consistency.Offers Closed System: Gives the ability to assemble in Open Lab type EnvironmentProvides aseptic solution to close off an unused line.How to Sterilize: Autoclave Max 15 psi @ 134 degreesC, 60 minGamma Radiation 25 kGy to 50 kGy.