Learn More
Andwin Scientific Tri-Corn Beakers, 250ml, 100/pack
SDP
Catalog No. 50996332
Encompass_Preferred
The Tri-Corn Beakers feature three dripless pouring spouts. This product is resistant to most chemicals and is autoclavable. It can resist temperatures up to 121C. The beakers have volume capacity of 250 mL and has graduations at 10 mL. This is sold in a pack of 100.
Catalog No. 50-996-332 Supplier Andwin Scientific Supplier No. 60978
The Tri-Corn Beakers feature three dripless pouring spouts. This product is resistant to most chemicals and is autoclavable. It can resist temperatures up to 121C. The beakers have volume capacity of 250 mL and has graduations at 10 mL. This is sold in a pack of 100.