Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Plastic Disposable Tri-Corn Beaker, 100 ml, 10 ml gradations, 73 mm height
SDP

Catalog No. 501898984
Encompass_Preferred

Three dripless pouring spouts. Resistant to most chemicals. Autoclavable. The beakers can withstand temperatures up to 121 degree C. Metric graduations.
Capacity: 100 ml
Gradations: 10 ml
Height: 73 mm
Diameter: 53 mm

Catalog No. 50-189-8984 Supplier Electron Microscopy Sciences Supplier No. 60970
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Three dripless pouring spouts. Resistant to most chemicals. Autoclavable. The beakers can withstand temperatures up to 121 degree C. Metric graduations.
Capacity: 100 ml
Gradations: 10 ml
Height: 73 mm
Diameter: 53 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.