Learn More
Andwin Scientific TRI-POUR BEAKER 400ML 100/PKT
SDP
Catalog No. 50996328
Encompass_Preferred
The Tri-Corn Beakers feature three dripless pouring spouts. This product is resistant to most chemicals and is autoclavable. It can resist temperatures up to 121C. The beakers have volume capacity of 400 mL and has graduations at 20 mL. This is sold in a pack of 100.
Catalog No. 50-996-328 Supplier Andwin Scientific Supplier No. 60974
Add to Cart
Add to Cart
The Tri-Corn Beakers feature three dripless pouring spouts. This product is resistant to most chemicals and is autoclavable. It can resist temperatures up to 121C. The beakers have volume capacity of 400 mL and has graduations at 20 mL. This is sold in a pack of 100.