Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Plastic Disposable Tri-Corn Beaker, 1000 ml, 50 ml gradations, 147 mm height
SDP

Catalog No. 501898990
Encompass_Preferred

Three dripless pouring spouts. Resistant to most chemicals. Autoclavable. The beakers can withstand temperatures up to 121 degree C. Metric graduations.
Capacity: 1000 ml
Gradations: 50 ml
Height: 147 mm
Diameter: 112 mm

Catalog No. 50-189-8990 Supplier Electron Microscopy Sciences Supplier No. 60978
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Three dripless pouring spouts. Resistant to most chemicals. Autoclavable. The beakers can withstand temperatures up to 121 degree C. Metric graduations.
Capacity: 1000 ml
Gradations: 50 ml
Height: 147 mm
Diameter: 112 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.