Learn More
National Optical Tri Power (1X/2X/4X) Stereo Microscope
SCI_ED
Catalog No. S81787
Change view
Click to view available options
Magnification Power:
1X, 2X, 4X
Catalog No. Magnification Power
S81787 1X, 2X, 4X
1 options
Catalog No. S81787 Supplier National Optical Supplier No. DELS1
Add to Cart
Add to Cart
The D-ELS-1 tri power stereo microscope features 10X, 20X, and 40X magnification and is ideal for elementary, middle, and high school education levels.
The D-ELS-1 tri power stereo microscope features 10X, 20X, and 40X magnification and is ideal for elementary, middle, and high school education levels. 
This budget-friendly stereo microscope features rechargeable top/bottom LED illumination, fixed arm, and stage clips.
MAGNIFICATION
  • 10X, 20X, and 40X Tri Power
FRAME
  • Fixed arm stand
ILLUMINATION
  • Top/Bottom LED Illumination rated up to 50,000 hours
  • Rechargeable
  • Batteries allow up to 500 recharges
STAGE
  • Locked-on stage clips
Specifications
Voltage 110/220 V
Magnification Power 1X, 2X, 4X
Product Type Stereo Microscope
Model D-ELS-1
Head Style Binocular
Lighting Type LED
Eyepiece Paired WF 10X
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov