Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tri(propylene glycol) methyl ether, mixture of isomers

Catalog No. 3028643L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
3 L
18 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-286-43L 3 L
30-286-418L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-286-43L Supplier Sigma Aldrich Supplier No. 3028643L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Tri(propylene glycol) methyl ether is a promising oxygenating agent for diesel fuels
  • Bubble coalescence and post-rupture oscillation in tri(propylene glycol) methyl ether solutions using high-speed cinematography was studied

Specifications

CAS 25498-49-1
Density 0.968 g/mL (at 25°C)
Boiling Point 100°C (2 mmHg)
Molecular Formula C10H22O4
Refractive Index n20/D 1.43 (literature)
Linear Formula CH3(OC3H6)3 OH
Quantity 3 L
Molecular Weight (g/mol) 206.28
Percent Purity 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.