Learn More
Molecular Research Center TRI REAGENT 100 ML
SDP
Catalog No. NC9387421
Encompass
Tri Reagent;; 100mL; For the single-step isolation/simultaneous of total RNA, DNA and proteins from tissues; cells cultured in monolayer or cell pellets1mL; 100mg tissue or 107 cells; Stable under normal temperatures and pressure
Catalog No. NC9387421 Supplier Molecular Research Center Supplier No. TR 118100ML
Tri Reagent;; 100mL; For the single-step isolation/simultaneous of total RNA, DNA and proteins from tissues; cells cultured in monolayer or cell pellets1mL; 100mg tissue or 107 cells; Stable under normal temperatures and pressure