Learn More
Molecular Research Center TRI REAGENT 200ML
SDP
Catalog No. NC9330796
Encompass
TRI Reagent ,200 ml ,reagent for the single-step isolation of total RNA or simultaneous isolation of RNA,DNA and proteins from tissues,cells cultured in monolayer or cell pellets.1 ml of TRI Reagent processes up to 100 mg tissue or 107 cell
Catalog No. NC9330796 Supplier Molecular Research Center Supplier No. TR 118200ML
TRI Reagent ,200 ml ,reagent for the single-step isolation of total RNA or simultaneous isolation of RNA,DNA and proteins from tissues,cells cultured in monolayer or cell pellets.1 ml of TRI Reagent processes up to 100 mg tissue or 107 cell