Learn More
Molecular Research Center TRI REAGENT 500 ML
SDP
Catalog No. NC9277980
Encompass
Reagent, Tri; Molecular Reaearch Centre; For RNA isolation; Isolation time: Less than one hour; 50mm sufficient to process 50 samples; 50 to 100mg of tissue each; 500mL
Catalog No. NC9277980 Supplier Molecular Research Center Supplier No. TR 118500ML
Reagent, Tri; Molecular Reaearch Centre; For RNA isolation; Isolation time: Less than one hour; 50mm sufficient to process 50 samples; 50 to 100mg of tissue each; 500mL