Learn More
Molecular Research Center TRI REAGENT 50ML
SDP
Catalog No. NC9324361
Encompass
TRI Reagent ,50 ml,reagent for the single-step isolation of total RNA or simultaneous isolation of RNA,DNA and proteins from tissues,cells cultured in monolayer or cell pellets.1 ml of TRI Reagent processes up to 100 mg tissue or 107 cells
Catalog No. NC9324361 Supplier Molecular Research Center Supplier No. TR 11850ML
TRI Reagent ,50 ml,reagent for the single-step isolation of total RNA or simultaneous isolation of RNA,DNA and proteins from tissues,cells cultured in monolayer or cell pellets.1 ml of TRI Reagent processes up to 100 mg tissue or 107 cells