Learn More
Molecular Research Center TRI REAGENT BD 100ML
SDP
Catalog No. NC9840025
Encompass
For single-step isolation of total RNA or simultaneous isolation of RNA, DNA and proteins from whole blood, plasma, or serum. 0.75 ml of TRI Reagent BD processes 0.25 ml of blood derivatives.
Catalog No. NC9840025 Supplier Molecular Research Center Supplier No. TB 126100ML
For single-step isolation of total RNA or simultaneous isolation of RNA, DNA and proteins from whole blood, plasma, or serum. 0.75 ml of TRI Reagent BD processes 0.25 ml of blood derivatives.