Learn More
Molecular Research Center TRI REAGENT LS 200 ML
SDP
Catalog No. NC9574779
Encompass
For single-step isolation of total RNA or simultaneous isolation of RNA, DNA and proteins from liquid samples. 0.75 ml of TRI Reagent LS processes 0.25 ml of liquid samples.
Catalog No. NC9574779 Supplier Molecular Research Center Supplier No. TS 120200ML
For single-step isolation of total RNA or simultaneous isolation of RNA, DNA and proteins from liquid samples. 0.75 ml of TRI Reagent LS processes 0.25 ml of liquid samples.