Learn More
Molecular Research Center TRI REAGENT LS 50ML
SDP
Catalog No. NC9000548
Encompass
TRI Reagent LS, A reagent for the single-step isolation of total RNA or simultaneous isolation of RNA, DNA and proteins from liquid samples such as cell suspensions, Serum, CSF and amniotic fluid, 50mL
Catalog No. NC9000548 Supplier Molecular Research Center Supplier No. TS 12050ML
TRI Reagent LS, A reagent for the single-step isolation of total RNA or simultaneous isolation of RNA, DNA and proteins from liquid samples such as cell suspensions, Serum, CSF and amniotic fluid, 50mL