Learn More
Molecular Research Center TRI REAGENT RT 100ML
SDP
Catalog No. NC9006102
Encompass
A reagent for the single-step isolation of RNA from tissues and cells. 1 ml of TRI Reagent RT processes 50 mg tissue or 107 cells. The isolated RNA is free of DNA contamination and ready for RT-PCR without the DNase treatment.
Catalog No. NC9006102 Supplier Molecular Research Center Supplier No. RT 111100ML
A reagent for the single-step isolation of RNA from tissues and cells. 1 ml of TRI Reagent RT processes 50 mg tissue or 107 cells. The isolated RNA is free of DNA contamination and ready for RT-PCR without the DNase treatment.