Learn More
Molecular Research Center TRI Reagent RT, 200 mL
SDP
Catalog No. NC0850610
Encompass
TRI Reagent RT is used for RNA isolation from tissues, pelleted cells and cells grown in monolayer. Fifty milliliters is sufficient to process 50 samples, each containing 50 mg of tissue. Expected yields range from 50 – 700 ug of RNA per sample, depending upon the tissue source.
Catalog No. NC0850610 Supplier Molecular Research Center Supplier No. RT 111200ML
TRI Reagent RT is used for RNA isolation from tissues, pelleted cells and cells grown in monolayer. Fifty milliliters is sufficient to process 50 samples, each containing 50 mg of tissue. Expected yields range from 50 – 700 ug of RNA per sample, depending upon the tissue source.