Learn More
Molecular Research Center TRI REAGENT RT 50ML
SDP
Catalog No. NC9785622
Encompass
A reagent for the single-step isolation of RNA from tissues and cells. 1 ml of TRI Reagent RT processes 50 mg tissue or 107 cells. The isolated RNA is free of DNA contamination and ready for RT-PCR without the DNase treatment.
Catalog No. NC9785622 Supplier Molecular Research Center Supplier No. RT 11150ML
A reagent for the single-step isolation of RNA from tissues and cells. 1 ml of TRI Reagent RT processes 50 mg tissue or 107 cells. The isolated RNA is free of DNA contamination and ready for RT-PCR without the DNase treatment.