Learn More
Electron Microscopy Sciences Tri-Seal™ Triple Seal Specimen Container, 240 mL
SDP
Supplier:  Electron Microscopy Sciences 64235240
Encompass_Preferred
Leakproof, airtight, touch-top specimen containers for the pathology lab. No more loose threaded screw caps to deal with – just press down the triple-seal lid to securely close and seal.
Volume: 240 mL
Catalog No. 50-334-68