Learn More
Electron Microscopy Sciences Tri-Seal™ Triple Seal Specimen Container Tall with Locking Latch, 50 mL
SDP
Catalog No. 5033470
Encompass_Preferred
Leakproof, airtight, touch-top specimen containers for the pathology lab. No more loose threaded screw caps to deal with – just press down the triple-seal lid to securely close and seal.
Volume: 50 mL
Catalog No. 50-334-70 Supplier Electron Microscopy Sciences Supplier No. 6423550TL
Add to Cart
Add to Cart
Leakproof, airtight, touch-top specimen containers for the pathology lab. No more loose threaded screw caps to deal with – just press down the triple-seal lid to securely close and seal.
Volume: 50 mL