Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tri-Sodium Citrate Dihydrate, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1064485000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
5 kg
25 kg
Packaging:
Fiber Carton
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1064485000 5 kg Plastic Bottle
M1064480500 500 g Plastic Bottle
M1064489025 25 kg Fiber Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1064485000 Supplier MilliporeSigma Supplier No. 1064485000
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4-3-6132
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]

Specifications

Melting Point 150°C (anhydrous substance)
Color White
pH 7.5 to 9.0 (50g/L, H2O, 25°C)
Physical Form Powder
Quantity 5 kg
Solubility Information 720g/L
Formula Weight 294.1
Percent Purity 99.0 to 101.0%
Grade ACS
Packaging Plastic Bottle
Chemical Name or Material Sodium Citrate Dihydrate
Show More Show Less
EINECSNumber 200-675-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.