Learn More
Strem, An Ascensus Company CAS# 131274-22-1. 1g. Tri-t-butylphosphonium tetrafluoroborate, 99%. MFCD04039975
SDP
Catalog No. 5090110380
Encompass
CAS# 131274-22-1. 1g. Tri-t-butylphosphonium tetrafluoroborate, 99%. MFCD04039975. Molecular Weight: 290.13. Molecular Formula: [(C4H9)3PH]+BF4ae. Color/form: white pwdr. Strem# 15-6000. http://www.strem.com/catalog/v/15-6000/
Catalog No. 50-901-10380 Supplier Strem, An Ascensus Company Supplier No. 1560001G
CAS# 131274-22-1. 1g. Tri-t-butylphosphonium tetrafluoroborate, 99%. MFCD04039975. Molecular Weight: 290.13. Molecular Formula: [(C4H9)3PH]+BF4ae. Color/form: white pwdr. Strem# 15-6000. http://www.strem.com/catalog/v/15-6000/