Learn More
Pharmacal Research Laboratories TRI TEMPS STICKERS 16/PK
SDP
Catalog No. NC0292506
Encompass
Tri-Temp Tapes are non-reversible, cage surface temperature-recording labels. The light gray backgrounds on each bar will turn black seconds after being exposed to 160o, 170o and 180o F water respectively. One tape shows all three levels. Accuracy of this tape is 99%. The exposed recording tape may be removed and filed for future reference. We recommend the use of this tape for verifying every load in all automatic cage washing equipment.
Catalog No. NC0292506 Supplier Pharmacal Research Laboratories Supplier No. 99064
Tri-Temp Tapes are non-reversible, cage surface temperature-recording labels. The light gray backgrounds on each bar will turn black seconds after being exposed to 160o, 170o and 180o F water respectively. One tape shows all three levels. Accuracy of this tape is 99%. The exposed recording tape may be removed and filed for future reference. We recommend the use of this tape for verifying every load in all automatic cage washing equipment.